missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, 10% w/v aq. soln.

Product Code. 15494749
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494749 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15494749 Supplier Thermo Scientific Alfa Aesar Supplier No. J62355.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol

Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, d
TRUSTED_SUSTAINABILITY

Specifications

CAS 76-03-9
Color Colorless
Concentration 10% w/v aq. soln.
Quantity 500 mL
Odor Acrid
Physical Form Liquid
Molecular Formula C2HCl3O2
Linear Formula Cl3CCO2H
MDL Number MFCD00004177
UN Number UN2564
Beilstein 970119
Merck Index 14,9627
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
Solubility Information Soluble in water (1200mg/ml at 20°C),alcohol,ether,ethanol (very soluble),and carbon tetrachloride.
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES C(=O)(C(Cl)(Cl)Cl)O
IUPAC Name 2,2,2-trichloroacetic acid
Molecular Weight (g/mol) 163.378
PubChem CID 6421
ChEBI CHEBI:30956
Formula Weight 163.39
Chemical Name or Material Trichloroacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trichloroacetic acid
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Trichloroacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.