missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, 99+%, ACS reagent

Product Code. 10065880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10065880 500 g Glass Bottle 500g
10391351 100 g Glass Bottle 100g
11964921 2.5 kg Glass Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10065880 Supplier Thermo Scientific Acros Supplier No. 421455000
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Color White
Density 1.6200g/mL
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Adhering Crystals
Assay Percent Range 99+%
Residue after Ignition 0.03% max.
Linear Formula CCl3CO2H
Vapor Pressure 1.2mbar at 50°C
Beilstein 02, 206
Fieser 01,1194; 02,425; 04,520; 15,168
Merck Index 15, 9792
Formula Weight 163.39
Percent Purity ≥99.0%
Grade ACS Reagent
Chemical Name or Material Trichloroacetic acid, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trichloroacetic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.