missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, 99%, extra pure

Product Code. 13488368 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
10kg
1kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13488368 10 kg Plastic Drum 10kg
10715991 500 g Plastic bottle 500g
10377560 1 kg Plastic bottle 1kg
10437903 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13488368 Supplier Thermo Scientific Acros Supplier No. 152130100
  • ONLINE EXCLUSIVE PRICE
    835.00 €
    810.00 € / 10kg
    Save 25.00 €
    2% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76-03-9 | C2HCl3O2 | 163.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.39
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Melting Point 52.0°C to 58.0°C
Boiling Point 196.0°C
Color White
Density 1.6200 g/mL
Quantity 10 kg
Infrared Spectrum Authentic
Packaging Plastic Drum
Physical Form Flakes, Crystals, or Crystalline Powder
Assay Percent Range 0.99
Specific Gravity 1.62
Linear Formula CCl3CO2H
Beilstein 2206
Fieser 01,1194; 02,425; 04,520; 15,168
Merck Index 159792
Solubility Information Solubility in water: 120g/100 mL (20°C). Other solubilities: soluble in alcohol and ether,carbon tetrachloride
Formula Weight 163.39
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Trichloroacetic acid, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.