missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™

Product Code. 10775151 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10775151 1 kg 1kg
10764961 250 g 250g
10162590 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10775151 Supplier Fisher Chemical Supplier No. T/3000/60
  • ONLINE EXCLUSIVE PRICE
    296.00 €
    165.00 € / 1kg
    Save 131.00 €
    44% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

C2HCl3O2, CAS Number-76-03-9, trichloressigsaeure, konesta, amchem grass killer, acido tricloroacetico, acide trichloracetique, aceto-caustin, acetic acid, trichloro, trichloroethanoic acid, trichloroacetic acid, trichloroacetic acid, 500g, 196 deg.C, CHEBI:30956, White, 163.39g/mol | CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol

C2HCl3O2, CAS Number-76-03-9, trichloressigsaeure, konesta, amchem grass killer, acido tricloroacetico, acide trichloracetique, aceto-caustin, acetic acid, trichloro, trichloroethanoic acid, trichloroacetic acid, trichloroacetic acid, 500g, 196 deg.C, CHEBI:30956, White, 163.39g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number 4177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

pH 1.2
Melting Point 52°C
Boiling Point 196°C
Color White
Formula Weight 163.39g/mol
Quantity 1 kg
Packaging Plastic powder jar
Physical Form Solid
Vapor Pressure 1.2mbar at 50°C, 0.08mbar @25C
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trichloroacetic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.