missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15567834 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
125 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
125g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15567834 125 g Wide Mouth Amber Glass Bottle 125g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15567834 Supplier Avantor J.T.Baker Supplier No. 041404
  • ONLINE EXCLUSIVE PRICE
    189.57 €
    170.09 € / 125g
    Save 19.48 €
    10% Off
 Restricted Item
Estimated Shipment: 15-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Formula Weight 163.39
Grade BAKER ANALYZED™ ACS Reagent
Quantity 125 g
Packaging Wide Mouth Amber Glass Bottle
Physical Form Crystalline
Specific Gravity 1L = 1.60kg
Chemical Name or Material Trichloroacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.