missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, MP Biomedicals™

Product Code. 11497912 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497912 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11497912 Supplier MP Biomedicals Supplier No. 0215259290
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used as a decalcifier and fixative in microscopy. Trichloroacetic Acid, MP Biomedicals is used in protein precipitation and has been used to determine protein concentration by quantitative precipitation. A protocol for the precipitation of nucleic acids c

TRUSTED_SUSTAINABILITY

Specifications

CAS 76-03-9
Molecular Formula C2HCl3O2
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
Solubility Information Soluble in water; very soluble in ethanol or ether. Decomposed by heating with caustic alkalies into chloroform and alkali carbonate.
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES C(=O)(C(Cl)(Cl)Cl)O
IUPAC Name 2,2,2-trichloroacetic acid
Molecular Weight (g/mol) 163.378
PubChem CID 6421
ChEBI CHEBI:30956
Formula Weight 163.4
Percent Purity ≥99.0%
Grade ACS
Quantity 500 g
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.