missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid (TCA), BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10385960 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10385960 250 g 250g
10549780 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10385960 Supplier JT Baker Supplier No. 0344.0250
  • ONLINE EXCLUSIVE PRICE
    92.23 €
    73.75 € / 250g
    Save 18.48 €
    20% Off
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Exceeds ACS Specifications

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.38
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym TCA
IUPAC Name trichloroacetic acid
SMILES OC(=O)C(Cl)(Cl)Cl

Specifications

Melting Point 54°C to 56°C
Boiling Point 196°C (1013 hPa)
Assay Percent Range min. 99.0%
Density 1.63 g/cm³ (20°C)
Quantity 250 g
Flash Point 113°C
Packaging Glass bottle for solids
Residue after Ignition max. 0.02%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.