missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, USP 21, 99-100.5% (calc. to dried subst), Solstice
BIOPHARMA

Product Code. 16645783
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16645783 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16645783 Supplier Fluka Supplier No. 27242H1KG
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Specifications

pH 1
CAS 76-03-9
Melting Point 54°C to 58°C
Boiling Point 196°C
CAS Max % 1
Color White
Density 1.63 g/cm3 (20°C)
Quantity 1 kg
Flash Point 113°C
Molecular Formula C2HCl3O2
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES OC(=O)C(Cl)(Cl)Cl
IUPAC Name trichloroacetic acid
Molecular Weight (g/mol) 163.38
Grade Reagent Pharmacopeia Acids
Chemical Name or Material Trichloroacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.