missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetyl isocyanate, NMR grade

Product Code. 10678314 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10678314 2 g Glass bottle 2g
10636232 10 g Glass bottle 10g
13498458 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10678314 Supplier Thermo Scientific Acros Supplier No. 202900020
  • ONLINE EXCLUSIVE PRICE
    44.75 €
    43.40 € / 2g
    Save 1.35 €
    3% Off
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3019-71-4 | C3Cl3NO2 | 188.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3019-71-4
Molecular Formula C3Cl3NO2
Molecular Weight (g/mol) 188.39
MDL Number MFCD00002033
InChI Key GRNOZCCBOFGDCL-UHFFFAOYSA-N
Synonym trichloroacetyl isocyanate, acetyl isocyanate, trichloro, trichloroacetylisocyanate, isocyanic acid trichloroacetyl ester, unii-qxv1p5j6jl, qxv1p5j6jl, acetyl isocyanate, 2,2,2-trichloro, 2,2,2-trichloroethanecarbonyl isocyanate, trichloroethanecarbonyl isocyanate, trichloroacetyl isocyanate, nmr grade
PubChem CID 76400
IUPAC Name 2,2,2-trichloroacetyl isocyanate
SMILES ClC(Cl)(Cl)C(=O)N=C=O

Specifications

Color Colorless to Yellow
Density 1.5900g/mL
Boiling Point 80.0°C to 85.0°C (20.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CCl3CONCO
Refractive Index 1.4799 to 1.4819
Quantity 2 g
Fieser 02,425; 07,382
Specific Gravity 1.59
Solubility Information Solubility in water: decomposes
Formula Weight 188.4
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Trichloroacetyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.