missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tricine, 99+%, for biochemistry

Product Code. 10021811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
25g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021811 100 g Plastic bottle 100g
10027882 25 g Plastic bottle 25g
10742325 1 kg Plastic bottle 1kg
10676184 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10021811 Supplier Thermo Scientific Chemicals Supplier No. 327831000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine, N-[Tris(hydroxymethyl)methyl]glycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Absorbance 0.02 max. at 280nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point 180.0°C to 184.0°C
Color White
pH 4.6 to 5.6 (1% aq. soln. at 25°C)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (HOCH2)3CNHCH2CO2H
Packaging Plastic bottle
Solubility Information Solubility in water: soluble.
Formula Weight 179.17
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material Tricine
Show More Show Less
Product Identifier
  • Tricine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.