missing translation for 'onlineSavingsMsg'
Learn More

Triclocarban, MedChemExpress

Product Code. 16452286
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452286 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452286 Supplier Medchem Express Supplier No. HYB1805
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Triclocarban (3,4,4′-Trichlorocarbanilide), a broad spectrum antibacterial compound, is widely used in a broad range of applications such as the production of soaps, skin creams, toothpastes and deodorants. Triclocarban is a potential endocrine-disrupting chemical with the capacity to modulate androgen and estrogen activities as well as other hormone-mediated biological processes.

Triclocarban (3,4,4′-Trichlorocarbanilide), a broad spectrum antibacterial compound, is widely used in a broad range of applications such as the production of soaps, skin creams, toothpastes and deodorants. Triclocarban is a potential endocrine-disrupting chemical with the capacity to modulate androgen and estrogen activities as well as other hormone-mediated biological processes.

Specifications

Chemical Name or Material Triclocarban
Physical Form Solid
CAS 101-20-2
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C13H9Cl3N2O
For Use With (Application) COVID-19-immunoregulation
Synonym 3,4,4′-Trichlorocarbanilide
Solubility Information DMSO : 100 mg/mL (316.88 mM; Need ultrasonic)
SMILES ClC1=CC=C(C=C1)NC(NC2=CC=C(C(Cl)=C2)Cl)=O
Molecular Weight (g/mol) 315.58
Formula Weight 315.58
Percent Purity 98.85%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.