missing translation for 'onlineSavingsMsg'
Learn More

Triclosan, MedChemExpress

Product Code. 16418626
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418626 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418626 Supplier Medchem Express Supplier No. HYB1119
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Triclosan is an antibacterial and antifungal agent found in consumer products, including soaps, detergents, toys, and surgical cleaning treatments.

Triclosan is an antibacterial and antifungal agent found in consumer products, including soaps, detergents, toys, and surgical cleaning treatments.

Specifications

Chemical Name or Material Triclosan
Physical Form Solid
CAS 3380-34-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H7Cl3O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (345.38 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=CC(Cl)=CC=C1OC2=CC=C(Cl)C=C2Cl
Molecular Weight (g/mol) 289.54
Formula Weight 289.54
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.