missing translation for 'onlineSavingsMsg'
Learn More

Triethylsulfonium iodide, 97%

Product Code. 11300164
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11300164 5 g 5g
11310164 25 g 25g
11320164 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11300164 Supplier Thermo Scientific Alfa Aesar Supplier No. L00667.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1829-92-1 | C6H15IS | 246.15 g/mol

Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Light, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1829-92-1
Molecular Formula C6H15IS
Molecular Weight (g/mol) 246.15
MDL Number MFCD00051867
InChI Key LKDQWVKWYGOVJW-UHFFFAOYSA-M
Synonym triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide
PubChem CID 74589
IUPAC Name triethylsulfanium;iodide
SMILES CC[S+](CC)CC.[I-]

Specifications

Melting Point ∽148°C (decomposition)
Quantity 5 g
Beilstein 3911790
Sensitivity Light sensitive
Solubility Information It is soluble in water.
Formula Weight 246.15
Percent Purity 97%
Chemical Name or Material Triethylsulfonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.