missing translation for 'onlineSavingsMsg'
Learn More

Trifloxystrobin (free acid), Dr. Ehrenstorfer

Product Code. 30326753
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326753 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326753 Supplier Dr. Ehrenstorfer Supplier No. DREC17842200
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trifloxystrobin (free acid)
CAS 252913-85-2
Analyte or Component Names Trifloxystrobin (free acid)
Shipping Condition Room Temperature
Molecular Formula C19 H17 F3 N2 O4
Certifications/Compliance ISO 17034
Synonym (αE)-α-(Methoxyimino)-2-[[[(E)-[1-[3-(trifluoromethyl)phenyl]ethylidene]amino]oxy]methyl]benzeneacetic acid, (E,E)-α-(Methoxyimino)-2-[([1-[3-(trifluoromethyl)phenyl]ethylidene]amino)oxymethyl]benzeneacetic acid, CGA 321113, Trifloxystrobin acid
InChI Formula InChI=1S/C19H17F3N2O4/c1-12(13-7-5-8-15(10-13)19(20,21)22)23-28-11-14-6-3-4-9-16(14)17(18(25)26)24-27-2/h3-10H,11H2,1-2H3,(H,25,26)/b23-12+,24-17+
Solution Type Neat
SMILES CO\N=C(\C(=O)O)/c1ccccc1CO\N=C(/C)\c2cccc(c2)C(F)(F)F
IUPAC Name (2E)-2-methoxyimino-2-[2-[[(E)-1-[3-(trifluoromethyl)phenyl]ethylideneamino]oxymethyl]phenyl]acetic acid
Industry Type Environmental Testing
Molecular Weight (g/mol) 394.34
Quantity 10 mg
Formula Weight 394.114 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.