missing translation for 'onlineSavingsMsg'
Learn More

Triflumuron, TRC

Product Code. 30138705
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138705 100 g 100g
30135281 25 g 25g
30040793 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138705 Supplier Toronto Research Chemicals Supplier No. TRCT779100100G
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64628-44-0
Molecular Formula C15 H10 Cl F3 N2 O3
Molecular Weight (g/mol) 358.7
Synonym Bay Sir 8514, 1-(2-chlorobenzoyl)-3-(4-trifluoromethoxyphenyl)urea
IUPAC Name 2-chloro-N-[[4-(trifluoromethoxy)phenyl]carbamoyl]benzamide
SMILES FC(F)(F)Oc1ccc(NC(=O)NC(=O)c2ccccc2Cl)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C15H10ClF3N2O3/c16-12-4-2-1-3-11(12)13(22)21-14(23)20-9-5-7-10(8-6-9)24-15(17,18)19/h1-8H,(H2,20,21,22,23)
Formula Weight 358.0332
Chemical Name or Material Triflumuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.