missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Trifluoro-m-cresol, 98+%

Product Code. 10164420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10164420 25 mL Glass bottle 25mL
10154470 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10164420 Supplier Thermo Scientific Acros Supplier No. 168470250
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-17-9 | C7H5F3O | 162.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-17-9
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.11
MDL Number MFCD00002299
InChI Key UGEJOEBBMPOJMT-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
PubChem CID 7376
IUPAC Name 3-(trifluoromethyl)phenol
SMILES C1=CC(=CC(=C1)O)C(F)(F)F

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -1.8°C
Color Yellow
Density 1.3300g/mL
Boiling Point 178°C to 179°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CF3C6H4OH
Refractive Index 1.4570 to 1.459
Quantity 25 mL
Beilstein 06, I, 187
Specific Gravity 1.33
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 162.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluoro-m-cresol, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a,a,a-Trifluoro-m-cresol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.