missing translation for 'onlineSavingsMsg'
Learn More

α,α,α-Trifluoro-m-tolyl isothiocyanate, 98%, ACROS Organics™

Product Code. 10013740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10013740 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10013740 Supplier Thermo Scientific Acros Supplier No. 306890050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1840-19-3
CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Orange
Density 1.3350g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H4F3NS
Linear Formula CF3C6H4NCS
Refractive Index 1.5550 to 1.5570 (20°C, 589nm)
MDL Number MFCD00004806
Quantity 5 g
Synonym 3-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl benzene, 3-trifluoromethyl phenylisothiocyanate, benzene, 1-isothiocyanato-3-trifluoromethyl, m-trifluoromethyl phenyl isothiocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isothiocyanate, 3-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl-benzene, benzene,1-isothiocyanato-3-trifluoromethyl
InChI Key GFEPANUKFYVALF-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)N=C=S)C(F)(F)F
IUPAC Name 1-isothiocyanato-3-(trifluoromethyl)benzene
Molecular Weight (g/mol) 203.182
PubChem CID 137232
Formula Weight 203.19
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluoro-m-tolyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.