missing translation for 'onlineSavingsMsg'
Learn More

(alpha,alpha,alpha-Trifluoro-p-tolyl)acetic acid, 98%

Product Code. 10361514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10361514 5 g Glass bottle 5g
10647182 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10361514 Supplier Thermo Scientific Acros Supplier No. 214370050
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 32857-62-8 | C9H7F3O2 | 204.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32857-62-8
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00004352
InChI Key HNORVZDAANCHAY-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylacetic acid, 2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylphenylacetic acid, 4-trifluoromethyl phenyl acetic acid, alpha,alpha,alpha-trifluoro-p-tolyl acetic acid, benzeneacetic acid, 4-trifluoromethyl, p-trifluoromethyl phenylacetic acid, p-trifluoromethylphenylacetic acid, alpha,alpha,alpha-trifluoro-4-tolyl acetic acid
PubChem CID 118342
IUPAC Name 2-[4-(trifluoromethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC=C(C=C1)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 81°C to 85°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CF3C6H4CH2CO2H
Quantity 5 g
Formula Weight 204.15
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (α, α, α-Trifluoro-p-tolyl)acetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.