missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoromethoxy)benzene, 99%

Product Code. 11451677
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451677 25 g 25g
11461677 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451677 Supplier Thermo Scientific Alfa Aesar Supplier No. B25658.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 456-55-3 | C7H5F3O | 162.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-55-3
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.11
MDL Number MFCD00040832
InChI Key GQHWSLKNULCZGI-UHFFFAOYSA-N
Synonym trifluoromethoxy benzene, 1-trifluoromethoxy benzene, benzene, trifluoromethoxy, phenyl trifluoromethyl ether, alpha,alpha,alpha-trifluoroanisole, trifluorophenoxymethane, pubchem7458, trifluoromethyloxybenzene, p-trifluoromethoxybenzene, trifluoromethoxy-benzene
PubChem CID 68010
IUPAC Name trifluoromethoxybenzene
SMILES FC(F)(F)OC1=CC=CC=C1

Specifications

Density 1.226
Boiling Point 102°C to 104°C
Flash Point 12°C (53°F)
Odor Aromatic
Refractive Index 1.406
Quantity 25 g
UN Number UN1993
Beilstein 2043132
Formula Weight 162.11
Percent Purity 99%
Chemical Name or Material (Trifluoromethoxy)benzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.