missing translation for 'onlineSavingsMsg'
Learn More

Trifluralin, Honeywell Fluka™

Product Code. 16654494 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16654494 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16654494 Supplier Honeywell Fluka Supplier No. R1774100MG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Trifluralin
CAS 1582-09-8
CAS Max % ≤ 100%
Linear Formula C13H16F3N3O4
Beilstein 1893555
Color Light Orange
Melting Point 47.2°C
Synonym 2,6-Dinitro-N,N-dipropyl-4-trifluoromethylaniline
Molecular Weight (g/mol) 335.28 g/mol
Quantity 100 mg
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.