missing translation for 'onlineSavingsMsg'
Learn More

Triformamide, 97%, Thermo Scientific™

Product Code. 10549403 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10549403 25 g Glass bottle 25g
10403253 1 g Glass bottle 1g
10691315 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10549403 Supplier Thermo Scientific Acros Supplier No. 380440250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 25891-31-0
MDL Number MFCD08059356
InChI Key PFBAGGWJGZAGCG-UHFFFAOYSA-N
Synonym triformamide
PubChem CID 6419719
IUPAC Name N,N-diformylformamide
SMILES C(=O)N(C=O)C=O

Specifications

Melting Point 95.0°C to 100.0°C
Color White
Elemental Analysis 96.0% min. (ex%C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3H3NO3
Linear Formula N(COH)3
Quantity 25 g
Molecular Weight (g/mol) 101.061
Formula Weight 101.06
Physical Form Crystals
Chemical Name or Material Triformamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.