missing translation for 'onlineSavingsMsg'
Learn More

Trihexyl Phosphate (>90%), TRC

Product Code. 30454515
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454515 500 mg 500mg
30449553 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454515 Supplier Toronto Research Chemicals Supplier No. TRCT794765500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2528-39-4
Molecular Formula C18H39O4P
Molecular Weight (g/mol) 350.47
Synonym Hexyl Phosphate, Tri(n-hexyl) Phosphate, Tri-N-hexyl Phosphate
IUPAC Name trihexyl phosphate
SMILES CCCCCCOP(=O)(OCCCCCC)OCCCCCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H39O4P/c1-4-7-10-13-16-20-23(19,21-17-14-11-8-5-2)22-18-15-12-9-6-3/h4-18H2,1-3H3
Formula Weight 350.2586
Chemical Name or Material Trihexyl Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.