missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM26 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | TRIM26 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TRIM26 Polyclonal antibody specifically detects TRIM26 in Human, Rat samples. It is validated for Western BlotSpecifications
| TRIM26 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 7726 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| Acid finger protein, AFP, RNF95RING finger protein 95, tripartite motif containing 26, tripartite motif-containing protein 26, widely expressed acid zinc finger protein, Zinc finger protein 173, ZNF173tripartite motif-containing 26 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TRIM26 (NP_001229712.1). MATSAPLRSLEEEVTCSICLDYLRDPVTIDCGHVFCRSCTTDVRPISGSRPVCPLCKKPFKKENIRPVWQLASLVENIERLKVDKGRQPGEVTREQQDAKLCERHREKLHYYCEDDGKLLCVMCRESREHRPHTAVLMEKAAQPHREKILNHLSTLRRDRDKIQGFQAKGEADILAALKKLQDQRQYIVAEFEQGHQFLREREEHLLEQLAKLEQELTEGREKFKSRGVGELARLALVISELEGKAQQPAAELMQDTRDFLNRYPRKKFWVGKPIARVVKKKTGEFSDKLLSLQRGLREF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title