missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM26 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94685-0.02ml
This item is not returnable.
View return policy
Description
TRIM26 Polyclonal antibody specifically detects TRIM26 in Human, Rat samples. It is validated for Western Blot
Specifications
| TRIM26 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| Acid finger protein, AFP, RNF95RING finger protein 95, tripartite motif containing 26, tripartite motif-containing protein 26, widely expressed acid zinc finger protein, Zinc finger protein 173, ZNF173tripartite motif-containing 26 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TRIM26 (NP_001229712.1). MATSAPLRSLEEEVTCSICLDYLRDPVTIDCGHVFCRSCTTDVRPISGSRPVCPLCKKPFKKENIRPVWQLASLVENIERLKVDKGRQPGEVTREQQDAKLCERHREKLHYYCEDDGKLLCVMCRESREHRPHTAVLMEKAAQPHREKILNHLSTLRRDRDKIQGFQAKGEADILAALKKLQDQRQYIVAEFEQGHQFLREREEHLLEQLAKLEQELTEGREKFKSRGVGELARLALVISELEGKAQQPAAELMQDTRDFLNRYPRKKFWVGKPIARVVKKKTGEFSDKLLSLQRGLREF | |
| 0.02 mL | |
| Zinc Finger | |
| 7726 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction