missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM44 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94800-0.1ml
This item is not returnable.
View return policy
Description
TRIM44 Polyclonal antibody specifically detects TRIM44 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TRIM44 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DIPBMGC3490, HSA249128, MC7, Protein DIPB, tripartite motif containing 44, tripartite motif-containing 44, tripartite motif-containing protein 44 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 205-344 of human TRIM44 (NP_060053.2). GAHQGHQLSTLDEAFEELRSKDSGGLKAAMIELVERLKFKSSDPKVTRDQMKMFIQQEFKKVQKVIADEEQKALHLVDIQEAMATAHVTEILADIQSHMDRLMTQMAQAKEQLDTSNESAEPKAEGDEEGPSGASEEEDT | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54765 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction