missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM49 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20721
This item is not returnable.
View return policy
Description
TRIM49 Polyclonal specifically detects TRIM49 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TRIM49 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| RING finger protein 18TRIM49L2, RNF18testis-specific RING Finger protein, Testis-specific RING-finger protein, tripartite motif containing 49, tripartite motif-containing 49, tripartite motif-containing protein 49 | |
| Rabbit | |
| 53 kDa | |
| 0.1 mL | |
| Zinc Finger | |
| 57093 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.68mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P0CI25 | |
| TRIM49 | |
| Recombinant protein encompassing a sequence within the center region of human TRIM49. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction