Learn More
TRIM56 Antibody (S02-8A3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19795-100ul
 View more versions of this product
Product Code. 18696053
  • 645.00 € / 100µL
Description
TRIM56 Monoclonal antibody specifically detects TRIM56 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRIM56
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunofluorescence
S02-8A3
Unconjugated
DKFZp667O116, E3 ubiquitin-protein ligase TRIM56, EC 6.3.2.-, RING finger protein 109, RNF109FLJ35608, tripartite motif containing 56, tripartite motif-containing 56, Tripartite motif-containing protein 56
A synthetic peptide of human TRIM56 (Uniprot # Q9BRZ2)
100 μg
Zinc Finger
81844
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.