missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TRIM72 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
- A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human TRIM72
- This peptide is useful as a blocking peptide for NBP1-86000.
- Protein was purified by IMAC chromatography.
- The expected concentration is greater than 0.5 mg/mL
- This product is intended to be used as a blocking antigen for antibody competition assays. Any other use of this antigen is done at the risk of the user. The use of this product for commercial production is strictly prohibited. Please contact technical support if you have any questions.
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| Gene ID (Entrez) | 493829 |
| Purification Method | >80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Antibody Competition |
| Gene Symbol | TRIM72 |
| Molecular Weight (g/mol) | 26 kDa |
| Quantity | 100 μL |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54960. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
| Storage Buffer | PBS and 1M Urea, pH 7.4. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction