Learn More
TRIM9 Monoclonal Antibody (OTI3C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800062
Encompass_Preferred
 View more versions of this product
Product Code. 17517482
  • 540.87 € / 100µL
Description
TRIM9 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.
Specifications
Specifications
TRIM9
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9C026
TRIM9
Human recombit protein fragment corresponding to amino acids 284-669 of human TRIM9 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3C9
Unconjugated
TRIM9
RNF91, SPRING
Mouse
Affinity Chromatography
RUO
114088
-20° C, Avoid Freeze/Thaw Cycles