missing translation for 'onlineSavingsMsg'
Learn More

Trimesitylphosphine, 98%

Product Code. 11484587
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11484587

Brand: Thermo Scientific Alfa Aesar B21116.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23897-15-6 | C27H33P | 388.535 g/mol

It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23897-15-6
Molecular Formula C27H33P
Molecular Weight (g/mol) 388.535
MDL Number MFCD00014912
InChI Key IDXDWPWXHTXJMZ-UHFFFAOYSA-N
Synonym trimesitylphosphine, tris 2,4,6-trimethylphenyl phosphine, tris 2,4,6-trimethylphenyl phosphane, phosphine, tris 2,4,6-trimethylphenyl, pubchem6447, acmc-1cs30, phosphine,tris 2,4,6-trimethylphenyl
PubChem CID 2733417
IUPAC Name tris(2,4,6-trimethylphenyl)phosphane
SMILES CC1=CC(=C(C(=C1)C)P(C2=C(C=C(C=C2C)C)C)C3=C(C=C(C=C3C)C)C)C

Specifications

Melting Point 185°C to 188°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 388.54
Percent Purity 98%
Chemical Name or Material Trimesitylphosphine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.