missing translation for 'onlineSavingsMsg'
Learn More

Trimethadione, MedChemExpress

Product Code. 16494177
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16494177 Supplier Medchem Express Supplier No. HYA0092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trimethadione (3,5,5,-Trimethyloxazolidine-2,4-dione) is an oxazolidinedione anticonvulsant agent widely used against absences seizures. Trimethadione also is a T-type calcium channel blocker which has antihyperalgesic effects.

Trimethadione (3,5,5,-Trimethyloxazolidine-2,4-dione) is an oxazolidinedione anticonvulsant agent widely used against absences seizures. Trimethadione also is a T-type calcium channel blocker which has antihyperalgesic effects.

Specifications

Chemical Name or Material Trimethadione
Physical Form Solid
CAS 127-48-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H9NO3
Synonym 3,5,5,-Trimethyloxazolidine-2,4-dione
Solubility Information DMSO : ≥ 40 mg/mL (279.45 mM)
SMILES O=C(N1C)OC(C)(C)C1=O
Molecular Weight (g/mol) 143.14
Formula Weight 143.14
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.