missing translation for 'onlineSavingsMsg'
Learn More

Melford Trimethoprim

Product Code. 30018874
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018874 5 g 5g
30018873 25 g 25g
30018872 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018874 Supplier Melford Supplier No. T590005.0
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Trimethoprim is a bacteriostatic antibiotic that inhibits bacterial dihydrofolate reductase, an enzyme involved in the synthesis of tetrahydrofolic acid.

Trimethoprim is a bacteriostatic antibiotic that inhibits bacterial dihydrofolate reductase, an enzyme involved in the synthesis of tetrahydrofolic acid. This acid is crucial for the production of nucleic acids (DNA and RNA) in bacteria. By blocking this enzyme, trimethoprim disrupts the folate synthesis pathway, leading to the inhibition of bacterial growth. Trimethoprim is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its presence in the culture medium allows for the specific isolation of bacteria that lack resistance to trimethoprim.

Specifications

Chemical Name or Material Trimethoprim
CAS 738-70-5
Quantity 5 g
Molecular Formula C14H18N4O3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.