missing translation for 'onlineSavingsMsg'
Learn More

Melford Trimethoprim

Product Code. p-200087201
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 30018873

Brand: Melford T5900025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trimethoprim is a bacteriostatic antibiotic that inhibits bacterial dihydrofolate reductase, an enzyme involved in the synthesis of tetrahydrofolic acid.

Trimethoprim is a bacteriostatic antibiotic that inhibits bacterial dihydrofolate reductase, an enzyme involved in the synthesis of tetrahydrofolic acid. This acid is crucial for the production of nucleic acids (DNA and RNA) in bacteria. By blocking this enzyme, trimethoprim disrupts the folate synthesis pathway, leading to the inhibition of bacterial growth. Trimethoprim is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its presence in the culture medium allows for the specific isolation of bacteria that lack resistance to trimethoprim.

Specifications

Chemical Name or Material Trimethoprim
CAS 738-70-5
Quantity 25 g
Molecular Formula C14H18N4O3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.