missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim, 99.8%, MP Biomedicals™

Product Code. 11495482 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495482 5 g 5g
11485482 1 g 1g
11405492 25 g 25g
11415492 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11495482 Supplier MP Biomedicals Supplier No. 0219552705
  • ONLINE EXCLUSIVE PRICE
    150.00 €
    141.75 € / 5g
    Save 8.25 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14H18N4O3
Molecular Weight (g/mol) 290.323
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
Synonym 2,4-Diamino-5-[3,4,5-trimethoxy-benzyl]pyrimidine
PubChem CID 5578
ChEBI CHEBI:45924
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N

Specifications

Type Trimethoprim
Physical Form Powder
Color White
Quantity 5 g
Assay Percent Range 0.998
Formula Weight 290.3
Percent Purity 99.8%
Grade Not Available
Chemical Name or Material Trimethoprim
compliance-icons
Product Identifier
  • Trimethoprim
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Trimethoprim
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.