missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthobenzoate, 97%

Product Code. 10722411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10722411 50 g Glass bottle 50g
10184140 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10722411 Supplier Thermo Scientific Acros Supplier No. 172920500
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 707-07-3 | C10H14O3 | 182.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 707-07-3
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00008474
InChI Key IECKAVQTURBPON-UHFFFAOYSA-N
Synonym trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate
PubChem CID 69720
IUPAC Name trimethoxymethylbenzene
SMILES COC(C1=CC=CC=C1)(OC)OC

Specifications

Color Colorless
Density 1.0600g/mL
Boiling Point 87°C to 88°C (7.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(OCH3)3
Refractive Index 1.4880 to 1.49
Quantity 50 g
Fieser 13,327; 04,548; 13,195; 01,1256; 02,454; 04,565; 06,657; 07,414; 08,524; 09,512; 10,455; 13,301
Specific Gravity 1.06
Formula Weight 182.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Trimethyl orthobenzoate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.