missing translation for 'onlineSavingsMsg'
Learn More

Trimethylammonium Bicarbonate Buffer, For HPLC, Honeywell™ Fluka™

Product Code. 15638320 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 15638320

Brand: Honeywell-Fluka 17899100ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 34408-21-4
Color Colorless
For Use With (Application) HPLC
pH 8.4 to 8.6 (at 25°C)
Physical Form Liquid
Quantity 100 mL
Molecular Formula C4H11NO3
Linear Formula (CH3)3N.H2CO3
MDL Number MFCD10565651
Packaging Plastic bottle
UN Number NONH for all modes of transport
InChI Key ARGNFRQXCRRALH-UHFFFAOYSA-N
SMILES CN(C)C.C(=O)(O)O
IUPAC Name carbonic acid;N,N-dimethylmethanamine
Molecular Weight (g/mol) 121.136
PubChem CID 186184
Formula Weight 121.14g/mol
Chemical Name or Material Trimethylammonium Bicarbonate Buffer
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.