missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIP15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13415237
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13415237 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13415237 Supplier Invitrogen™ Supplier No. PA530723

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG7, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (99%), Xenopus laevis (97%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the beta subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIP15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene COPS2
Gene Accession No. P61201, P61202
Gene Alias AI315723; ALIEN; alien homolog; Alien protein; alien-like; alien-PA; alien-PB; alien-PD; alien-PE; BcDNA:LD10463; beta-casein; C85265; CASB; casein beta; CG9556; CG9556-PA; CG9556-PB; CG9556-PD; CG9556-PE; CH2; COP9; COP9 (constitutive photomorphogenic) homolog, subunit 2; COP9 (constitutive photomorphogenic) homolog, subunit 2 (Arabidopsis thaliana); COP9 complex homolog subunit 2; COP9 constitutive photomorphogenic homolog subunit 2; COP9 signalosome complex subunit 2; COP9 signalosome subunit 2; COP9 signalosome subunit 2 CSN2; COPS2; Csn2; Csnb; d-Alien; DCH2; Dmel\CG9556; Dmel_CG9556; drosophila COP9 signalosome homolog 2; JAB1-containing signalosome subunit 2; SD08021p; SGN2; signalosome subunit 2; thyroid receptor interacting protein 15; thyroid receptor-interacting protein 15; TR-interacting protein 15; TRIP15; TRIP-15; tyroid receptor interacting protein 15
Gene Symbols COPS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TRIP15. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12848, 9318
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.