missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl borate, 97%

Product Code. 15452008
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15452008 5 g 5g
15462008 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452008 Supplier Thermo Scientific Alfa Aesar Supplier No. H55686.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1095-03-0 | C18H15BO3 | 290.125 g/mol

Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

Solubility
Soluble in chloroform, methanol.

Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Store under inert atmosphere.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1095-03-0
Molecular Formula C18H15BO3
Molecular Weight (g/mol) 290.125
MDL Number MFCD00059011
InChI Key MDCWDBMBZLORER-UHFFFAOYSA-N
Synonym triphenoxyborane, phenyl borate, triphenoxyboron, boric acid, triphenyl ester, phenyl borate, pho 3 b, trifenylester kyseliny borite, boric acid h3bo3 , triphenyl ester, triphenyl ester boric acid, trifenylester kyseliny borite czech, boric acid triphenyl ester
PubChem CID 14182
IUPAC Name triphenyl borate
SMILES B(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3

Specifications

Melting Point 98°C to 101°C
Linear Formula (C6H5O)3B
Quantity 5 g
UN Number UN2811
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,methanol.
Formula Weight 290.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Triphenyl borate
compliance-icons compliance-icons
Product Identifier
  • Triphenyl borate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Triphenyl borate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.