missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl Phosphate, Pure, Fisher Chemical

Product Code. 10497840 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10497840 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10497840 Supplier Fisher Chemical Supplier No. T/3650/53
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Plastic powder jar
CAS 115-86-6
Vapor Pressure 1.3 hPa 193
Physical Form Solid
Molecular Formula C18H15O4P
MDL Number MFCD00003031
Color White
Melting Point 47°C
Boiling Point 370°C
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
IUPAC Name triphenyl phosphate
Molecular Weight (g/mol) 326.29
PubChem CID 8289
Quantity 500 g
ChEBI CHEBI:35033
Formula Weight 326.28g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.