missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphite, 97%

Product Code. 11462948
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462948 500 g 500g
11472948 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462948 Supplier Thermo Scientific Alfa Aesar Supplier No. A18662.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-02-0 | C18H15O3P | 310.289 g/mol

Used to convert alcohols to alkyl halides, as a peptide coupling agent, and, in combination with ozone, as a low-temperature source of singlet oxygen, and ligand for metal-catalyzed reactions. Also used as reagent in the preparation of mixed valent nickel diphosphine dithiolate bridged iron carbonyl phosphine complexes as models of [NiFe]-hydrogenase active site.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to convert alcohols to alkyl halides, as a peptide coupling agent, and, in combination with ozone, as a low-temperature source of singlet oxygen, and ligand for metal-catalyzed reactions. Also used as reagent in the preparation of mixed valent nickel diphosphine dithiolate bridged iron carbonyl phosphine complexes as models of [NiFe]-hydrogenase active site.

Solubility
Insoluble in water.

Notes
Air and Moisture Sensitive, store away form air and moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-02-0
Molecular Formula C18H15O3P
Molecular Weight (g/mol) 310.289
MDL Number MFCD00003032
InChI Key HVLLSGMXQDNUAL-UHFFFAOYSA-N
Synonym phosphorous acid, triphenyl ester, advance tpp, triphenoxyphosphine, phenyl phosphite, stabilizer p 36, trifenoxyfosfin, trifenylfosfit, phosclere t 36, efed, mellite 310
PubChem CID 7540
IUPAC Name triphenyl phosphite
SMILES C1=CC=C(C=C1)OP(OC2=CC=CC=C2)OC3=CC=CC=C3

Specifications

Melting Point 22°C to 24°C
Density 1.184
Boiling Point 360°C
Flash Point 146°C (295°F)
Odor Mild
Linear Formula (C6H5O)3P
Quantity 500 g
UN Number UN3077
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 310.29
Percent Purity 97%
Chemical Name or Material Triphenyl phosphite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triphenyl phosphite
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Triphenyl phosphite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.