missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl Phosphite, TRC

Product Code. 30450441
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450441 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450441 Supplier Toronto Research Chemicals Supplier No. TRCT8089951G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 101-02-0
Molecular Formula C18 H15 O3 P
Quantity 1 g
Synonym Phenyl phosphite ((C6H5O)3P) (6CI,7CI), ADK Stab TPP, Advance TPP, Antioxidant TPP, Antioxidant TPPi, Doverphos 10, Irgafos TPP, Irgastab CH 55, JP 30, JP 360, Mark TPP, Mellite 310, NSC 43789, NSC 62219, Phosclere T 36, Plastistab 2334, Sumilizer TPP-R, Sumilizer TTP-R, TP 1, TP 1 (plasticizer), TP-I, TPP, TPP (plasticizer), TPP-R, Tppi, Triphenoxyphosphine, Triphenyl phosphite, Tris(phenoxy)phosphine, Weston TPP, Phosphorous acid, triphenyl ester, Triphenyl Phosphite
InChI Formula InChI=1S/C18H15O3P/c1-4-10-16(11-5-1)19-22(20-17-12-6-2-7-13-17)21-18-14-8-3-9-15-18/h1-15H
SMILES O(P(Oc1ccccc1)Oc2ccccc2)c3ccccc3
IUPAC Name triphenyl phosphite
Molecular Weight (g/mol) 310.28
Formula Weight 310.0759
Chemical Name or Material Triphenyl Phosphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.