missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethanol, 97%

Product Code. 10226160 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 10226160

Brand: Thermo Scientific Acros 158910500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76-84-6 | C19H16O | 260.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-84-6
Molecular Formula C19H16O
Molecular Weight (g/mol) 260.34
MDL Number MFCD00004445,MFCD10565638
InChI Key LZTRCELOJRDYMQ-UHFFFAOYSA-N
Synonym triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl
PubChem CID 6457
IUPAC Name triphenylmethanol
SMILES OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 160°C to 164°C
Color White to Yellow
Boiling Point 360°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula (C6H5)3COH
Quantity 50 g
Beilstein 06,713
Merck Index 15,9912
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. h2so4 and glacial acetic acid,soluble in alcohol,ether and benzene
Formula Weight 260.32
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Triphenylmethanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.