missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethyl bromide, 98%

Product Code. 10736432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10736432 50 g Glass bottle 50g
10575481 10 g Glass bottle 10g
10157743 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10736432 Supplier Thermo Scientific Acros Supplier No. 140390500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 596-43-0 | C19H15Br | 323.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-43-0
Molecular Formula C19H15Br
Molecular Weight (g/mol) 323.22
MDL Number MFCD00000120
InChI Key NZHXEWZGTQSYJM-UHFFFAOYSA-N
Synonym bromotriphenylmethane, triphenylmethyl bromide, trityl bromide, triphenylbromomethane, bromodiphenylmethyl benzene, methane, bromotriphenyl, benzene, 1,1',1-bromomethylidyne tris, triphenylmethylbromide, bromomethanetriyl tribenzene, 2-bromo-1,1,1-triphenylethane
PubChem CID 11692
IUPAC Name [bromo(diphenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br

Specifications

Melting Point 151°C to 155°C
Color Yellow
Boiling Point 230°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C6H5)3CBr
Quantity 50 g
Beilstein 05, 704
Fieser 01,1254; 16,375
Solubility Information Solubility in water: insoluble
Formula Weight 323.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Triphenylmethyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • Triphenylmethyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.