missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethyl chloride, 98%

Product Code. 10656342 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10656342 100 g Plastic bottle 100g
10666342 500 g Plastic bottle 500g
10185723 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10656342 Supplier Thermo Scientific Acros Supplier No. 140401000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76-83-5 | C19H15Cl | 278.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-83-5
Molecular Formula C19H15Cl
Molecular Weight (g/mol) 278.78
MDL Number MFCD00000813,MFCD00284810
InChI Key JBWKIWSBJXDJDT-UHFFFAOYSA-N
Synonym trityl chloride, triphenylmethyl chloride, triphenylchloromethane, chlorotriphenylmethane, chloromethanetriyl tribenzene, triphenylmethylchloride, methane, chlorotriphenyl, triphenyl chloromethane, chlorodiphenylmethyl benzene, benzene, 1,1',1-chloromethylidyne tris
PubChem CID 6456
SMILES ClC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 110.0°C to 113.0°C
Color Beige-Yellow or White
Boiling Point 230.0°C to 235.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5)3CCl
Quantity 100 g
Beilstein 05, 700
Fieser 01,1254; 02,453; 04,565; 05,741; 06,656; 15,359
Merck Index 15, 9944
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene, dichloromethane and dmf
IUPAC Name [chloro(diphenyl)methyl]benzene
Formula Weight 278.77
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Triphenylmethyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triphenylmethyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.