missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethyl mercaptan, 98+%

Product Code. 11422687
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422687 5 g 5g
11432687 25 g 25g
11442687 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422687 Supplier Thermo Scientific Alfa Aesar Supplier No. A14888.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3695-77-0 | C19H16S | 276.397 g/mol

Triphenylmethyl mercaptan, is used as a reagent used for the introduction of thiol group. Triphenylmethanethiol is a reagent used for the mild introduction of the mercapto functionality. It is also used as a pharmaceutical intermediate; Chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethyl mercaptan, is used as a reagent used for the introduction of thiol group. Triphenylmethanethiol is a reagent used for the mild introduction of the mercapto functionality. It is also used as a pharmaceutical intermediate; Chemical synthesis.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3695-77-0
Molecular Formula C19H16S
Molecular Weight (g/mol) 276.397
MDL Number MFCD00004854
InChI Key JQZIKLPHXXBMCA-UHFFFAOYSA-N
Synonym triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl
PubChem CID 77281
IUPAC Name triphenylmethanethiol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S

Specifications

Melting Point 103°C to 108°C
Odor Stench
Quantity 5 g
Beilstein 2113168
Solubility Information It is insoluble in water.
Formula Weight 276.4
Percent Purity ≥98%
Chemical Name or Material Triphenylmethyl mercaptan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.