missing translation for 'onlineSavingsMsg'
Learn More

(Triphenylmethyl)triphenylphosphonium chloride, 98%, Thermo Scientific™

Product Code. 11436016
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436016 250 g 250g
11416016 10 g 10g
11426016 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436016 Supplier Thermo Scientific Alfa Aesar Supplier No. B22319.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 48234-64-6
Molecular Formula C37H30ClP
Molecular Weight (g/mol) 541.071
MDL Number MFCD00082244
InChI Key LZOHWIXYBMRNAP-UHFFFAOYSA-M
Synonym triphenylmethyl triphenylphosphonium chloride, triphenyl triphenylmethyl phosphonium chloride, phosphonium, triphenyl triphenylmethyl-, chloride, triphenyl trityl phosphanium chloride, triphenyl trityl phosphonium chloride, acmc-1ala8, triphenyl trityl phosphonium, trityltriphenylphosphonium chloride, trityl triphenylphosphonium chloride
PubChem CID 85420
IUPAC Name triphenyl(trityl)phosphanium;chloride
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)[P+](C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Cl-]

Specifications

Melting Point 244°C to 245°C
Quantity 250 g
Beilstein 389941
Formula Weight 541.07
Percent Purity 98%
Chemical Name or Material (Triphenylmethyl)triphenylphosphonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.