missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphinecopper(I) hydride hexamer, 96%

Product Code. 10619224 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10619224 5 g 5g
10036672 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10619224 Supplier Thermo Scientific Acros Supplier No. 382510050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33636-93-0 | C108H96Cu6P6 | 1961.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33636-93-0
Molecular Formula C108H96Cu6P6
Molecular Weight (g/mol) 1961.07
MDL Number MFCD00221518
InChI Key IZZFKTUYPRVQID-UHFFFAOYSA-T
Synonym stryker's reagent, triphenylphosphine copper hydride hexamer, triphenylphosphine-copper i hydride hexamer, unii-8x3r32m7xu, hydrido triphenylphosphine copper i hexamer, stryker's reagent mi, triphenylphosphine cuprous hydride, triphenylphosphine copper i hydride hexamer, copper, hexa-mu3-hydrohexakis triphenylphosphine hexa-, octahedro, cuprous hydride triphenylphosphine hexamer
PubChem CID 11982471
IUPAC Name copper;triphenylphosphane
SMILES [H+].[H+].[H+].[H+].[H+].[H+].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)[Cu]123[Cu]45([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]11([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]22([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]34([P+](C3=CC=CC=C3)(C3=CC=CC=C3)C3=CC=CC=C3)[Cu]512[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Red
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Merck Index 15, 8987
Solubility Information Solubility in water: reacts slowly.
Formula Weight 1961.07
Percent Purity 96%
Physical Form Crystals
Chemical Name or Material Triphenylphosphinecopper(I) hydride hexamer
compliance-icons
Product Identifier
  • Triphenylphosphinecopper(I) hydride hexamer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Cuprous hydride triphenylphosphine hexamer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.