missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphonium bromide, 99%

Product Code. 10592831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10592831 100 g Glass bottle 100g
10360994 5 g Glass bottle 5g
10422441 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10592831 Supplier Thermo Scientific Acros Supplier No. 269091000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6399-81-1 | C18H16BrP | 343.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6399-81-1
Molecular Formula C18H16BrP
Molecular Weight (g/mol) 343.20
MDL Number MFCD00035107
InChI Key CMSYDJVRTHCWFP-UHFFFAOYSA-N
Synonym triphenylphosphine hydrobromide, triphenylphosphonium bromide, phosphine, triphenyl-, hydrobromide, triphenylphosphane hydrobromide, triphenylphosphine bromine, pubchem13868, ph3p hbr, ph3p . hbr, unappfzuzwmopv-uhfffaoysa-n
PubChem CID 80811
IUPAC Name triphenylphosphane;hydrobromide
SMILES Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 196.0°C to 199.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 100 g
Fieser 15,353
Formula Weight 343.2
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Triphenylphosphonium bromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Triphenylphosphonium bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.