missing translation for 'onlineSavingsMsg'
Learn More

Triphosphopyridine nucleotide, disodium salt, 97+%

Product Code. 10277553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10277553 100 mg Glass bottle 100mg
10222812 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10277553 Supplier Thermo Scientific Acros Supplier No. 360561000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24292-60-2 | C21H26N7Na2O17P3 | 787.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24292-60-2
Molecular Formula C21H26N7Na2O17P3
Molecular Weight (g/mol) 787.37
MDL Number MFCD00065390
InChI Key WSDDJLMGYRLUKR-MWLGWILCNA-L
Synonym nadp disodium salt, beta-nicotinamide adenine dinucleotide phosphate disodium salt
PubChem CID 131851653
SMILES [Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97% min. (enzymatic,on dry subst.)
Packaging Glass bottle
Quantity 100 mg
Solubility Information (0.05% in water) Clear colorless to light yellow
IUPAC Name disodium 1-[(2R,3R,4S,5R)-5-({[({[(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-(hydrogen phosphonatooxy)-3-hydroxyoxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Formula Weight 787.4
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Triphosphopyridine nucleotide, disodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.