accessibility menu, dialog, popup

UserName

CAS RN 24292-60-2

N(R)(R)N(R)N(R)OOPPO(R)(R)(R)(S)NNOOOOOOOOHONH2NH2HOOHNaNaPHOOO

CAS RN 24292-60-2

IUPAC Name: 1-(5-{[({[5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)oxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl)-3-carbamoyl-1λ⁵-pyridin-1-ylium disodium
Synonyms: β-nadp disodium
Molecular Weight (g/mol): 789.39
Molecular Formula: C21H28N7Na2O17P3
InChi Key: WINXVPJGUQYDJA-UHFFFAOYNA-N
SMILES: [Na].[Na].NC(=O)C1=C[N+](=CC=C1)C1OC(COP([O-])(=O)OP(O)(=O)OCC2OC(C(OP(O)(O)=O)C2O)N2C=NC3=C(N)N=CN=C23)C(O)C1O
Molecular Weight (g/mol) 
  • (3)
  • (10)
  • (2)
Quantity 
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (10)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (10)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (10)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)